siRNA Details
Gene Symbol: Iltifb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054079.2 | NP_473420.2 | 5 | 530 | ||
GenBank mRNA | AJ249492.1 | - | 5 | 530 | ||
AK005228.1 | - | 2 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
