siRNA Details
Gene Symbol: Tpbpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026429.4 | NP_080705.1 | 2 | 157 | ||
GenBank mRNA | AK005433.1 | - | 2 | 157 | ||
AK018793.1 | - | 2 | 156 | |||
AK018794.1 | - | 2 | 155 | |||
AK018795.1 | - | 2 | 157 | |||
AK018798.1 | - | 2 | 157 | |||
AK018799.1 | - | 2 | 157 | |||
AK018804.1 | - | 2 | 158 | |||
AK018805.1 | - | 2 | 157 | |||
AK018808.1 | - | 2 | 158 | |||
AK018809.1 | - | 2 | 156 | |||
AK018810.1 | - | 2 | 157 | |||
AK018817.1 | - | 2 | 157 | |||
AK018820.1 | - | 2 | 157 | |||
AK018824.1 | - | 2 | 157 | |||
AK018827.1 | - | 2 | 156 | |||
AY034576.1 | - | 2 | 132 | |||
BC099441.1 | - | 2 | 142 | |||
BC100333.1 | - | 2 | 142 | 30787126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
