siRNA Details
Gene Symbol: Slc19a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276455.1 | NP_001263384.1 | 6 | 2357 | ||
NM_054087.3 | NP_473428.1 | 6 | 2471 | |||
GenBank mRNA | AF179403.1 | - | 6 | 2462 | ||
AF216204.1 | - | 6 | 2441 | |||
AF360396.1 | - | 6 | 2453 | |||
AF418986.1 | - | 6 | 2348 | |||
AK044825.1 | - | 6 | 2472 | |||
BC004021.1 | - | 2 | 1025 | |||
BC034659.1 | - | 6 | 2452 | 5359902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
