siRNA Details
Gene Symbol: Pop5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026398.4 | NP_080674.1 | 5 | 646 | ||
GenBank mRNA | AK005290.1 | - | 4 | 402 | ||
AK012421.1 | - | 4 | 396 | |||
AK012523.1 | - | 404 | ||||
AK013257.1 | - | 3 | 383 | |||
BC022670.1 | - | 5 | 670 | 4224973 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
