siRNA Details
Gene Symbol: Ube3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054093.2 | NP_473434.2 | 13 | 1619 | ||
GenBank mRNA | AF244362.3 | - | 12 | 1254 | ||
BC023956.1 | - | 13 | 1388 | 5321098 | ||
BC034059.1 | - | 7 | 623 | 4163877 | ||
BC096743.1 | - | 13 | 1642 | 30549276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
