siRNA Details
Gene Symbol: Pip4k2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054097.3 | NP_473438.1 | 2 | 365 | ||
GenBank mRNA | AB054591.1 | - | 2 | 308 | ||
AK140079.1 | - | 2 | 329 | |||
AK148299.1 | - | 2 | 325 | |||
AK182167.1 | - | 1 | 58 | |||
BC021383.1 | - | 2 | 298 | 5042933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
