siRNA Details
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039511.1 | NP_001034600.1 | 6 | 948 | ||
NM_001039512.1 | NP_001034601.1 | 6 | 948 | |||
NM_054102.2 | NP_473443.2 | 6 | 948 | |||
GenBank mRNA | AB055737.1 | - | 6 | 944 | ||
AB055738.1 | - | 6 | 944 | |||
AK007291.1 | - | 6 | 898 | |||
AK129229.1 | - | 6 | 886 | |||
AK144619.1 | - | 6 | 927 | |||
AK144835.1 | - | 6 | 929 | |||
AK145071.1 | - | 6 | 939 | |||
AK145845.1 | - | 6 | 898 | |||
AK146001.1 | - | 6 | 929 | |||
AK146329.1 | - | 3 | 272 | |||
AK146494.1 | - | 6 | 901 | |||
AK146645.1 | - | 6 | 899 | |||
AK159254.1 | - | 6 | 930 | |||
BC004092.1 | - | 5 | 648 | 3593827 | ||
BC040250.1 | - | 6 | 940 | |||
DQ914522.1 | - | 6 | 900 | |||
DQ914523.1 | - | 6 | 870 |

More Information
Other Species Targeted

Gene Ontology Categories:
