siRNA Details
Gene Symbol: A1bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081067.1 | NP_001074536.1 | 6 | 1137 | ||
GenBank mRNA | BC080745.1 | - | 895 | |||
DQ520794.1 | - | 6 | 1133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
