siRNA Details
Gene Symbol: Gjc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080454.4 | NP_536702.3 | 2 | 913 | ||
NM_175452.4 | NP_780661.2 | 2 | 931 | |||
GenBank mRNA | AK046421.1 | - | 2 | 932 | ||
AK046609.1 | - | 2 | 914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
