siRNA Details
Gene Symbol: Mrps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080456.1 | NP_536704.1 | 3 | 428 | ||
GenBank mRNA | AB049943.1 | - | 3 | 443 | ||
AK165590.1 | - | 3 | 445 | |||
AK166374.1 | - | 3 | 443 | |||
AK167304.1 | - | 3 | 443 | |||
AK167316.1 | - | 3 | 443 | |||
AY061856.1 | - | 3 | 443 | |||
BC027271.1 | - | 3 | 428 | 3156971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
