siRNA Details
Gene Symbol: AF251705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134158.1 | NP_598919.1 | 2 | 520 | ||
GenBank mRNA | AB091767.1 | - | 2 | 251 | ||
AB091768.1 | - | 2 | 234 | |||
AB098476.1 | - | 2 | 187 | |||
AF251705.1 | - | 2 | 607 | |||
AI060397.1 | - | 2 | 276 | |||
AK050008.1 | - | 2 | 248 | |||
AK079395.1 | - | 2 | 336 | |||
AK154808.1 | - | 2 | 240 | |||
AY048685.1 | - | 2 | 520 | |||
AY457050.1 | - | 2 | 187 | |||
BC006801.1 | - | 2 | 251 | 3152857 | ||
BC111820.1 | - | 2 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
