siRNA Details
Gene Symbol: Emid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080595.2 | NP_542162.1 | 3 | 229 | ||
GenBank mRNA | AJ416093.1 | - | 3 | 229 | ||
AK153702.1 | - | 3 | 248 | |||
BC130023.1 | - | 3 | 234 | 40140754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
