siRNA Details
Gene Symbol: Gpr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199058.1 | NP_951013.1 | 1 | 484 | ||
GenBank mRNA | AK139367.1 | - | 2 | 663 | ||
AY064488.1 | - | 1 | 484 | |||
BC111861.1 | - | 1 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
