siRNA Details
Gene Symbol: Rem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080726.3 | NP_542764.2 | 3 | 668 | ||
GenBank mRNA | AK077317.1 | - | 3 | 549 | ||
AK090165.1 | - | 3 | 669 | |||
BC018219.1 | - | 3 | 550 | 3155806 |

More Information
Other Species Targeted

Gene Ontology Categories:
