siRNA Details
Gene Symbol: Tmprss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163776.1 | NP_001157248.1 | 8 | 850 | ||
NM_080727.2 | NP_542765.2 | 7 | 710 | |||
GenBank mRNA | AJ300738.1 | - | 7 | 710 | ||
AK158203.1 | - | 8 | 853 | |||
BC112375.1 | - | 7 | 707 | 40057032 |
