siRNA Details
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080849.3 | NP_543125.1 | 3 | 464 | ||
GenBank mRNA | AF407579.1 | - | 3 | 414 | ||
AK014546.1 | - | 3 | 425 | |||
AK154358.1 | - | 3 | 427 | |||
BB646522.1 | - | 3 | 474 | |||
BC057995.1 | - | 3 | 409 | |||
BC070457.1 | - | 3 | 413 | 30603080 |
