siRNA Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080853.3 | NP_543129.3 | 6 | 1503 | ||
GenBank mRNA | AF324864.1 | - | 6 | 1336 | ||
AK043753.1 | - | 5 | 956 | |||
AK045409.1 | - | 6 | 1266 | |||
BC038375.1 | - | 6 | 882 | 5357143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
