siRNA Details
Gene Symbol: Cntnap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130457.2 | NP_569724.2 | 17 | 2708 | ||
GenBank mRNA | AF333770.1 | - | 17 | 2673 | ||
AK081891.1 | - | 17 | 2701 | |||
AK142816.1 | - | 17 | 2704 | |||
AK173255.1 | - | 17 | 2683 | |||
BC034628.1 | - | 17 | 2708 | 5369648 |
