siRNA Details
Gene Symbol: Cntnap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130457.2 | NP_569724.2 | 4 | 606 | ||
GenBank mRNA | AF333770.1 | - | 4 | 571 | ||
AK042607.1 | - | 4 | 595 | |||
AK053611.1 | - | 4 | 598 | |||
AK081891.1 | - | 4 | 599 | |||
AK142816.1 | - | 4 | 602 | |||
AK173255.1 | - | 4 | 581 | |||
BC034628.1 | - | 4 | 606 | 5369648 |
