siRNA Details
Gene Symbol: Ubn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026666.3 | NP_080942.1 | 5 | 1279 | ||
GenBank mRNA | AK019173.1 | - | 4 | 1177 | ||
AK147571.1 | - | 5 | 1280 | |||
BC060723.1 | - | 3 | 207 | |||
BC098372.1 | - | 3 | 439 | 30363389 |

More Information
Other Species Targeted

Gene Ontology Categories:
