siRNA Details
Gene Symbol: Ubn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026666.3 | NP_080942.1 | 5, 6 | 1292 | ||
GenBank mRNA | AK019173.1 | - | 4, 5 | 1190 | ||
AK147571.1 | - | 5, 6 | 1293 | |||
BC060723.1 | - | 3, 4 | 220 | |||
BC098372.1 | - | 3, 4 | 452 | 30363389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
