siRNA Details
Gene Symbol: Krtap19-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130870.1 | NP_570940.1 | 1 | 555 | ||
GenBank mRNA | AF345291.1 | - | 1 | 555 | ||
AK137740.1 | - | 1 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
