siRNA Details
Gene Symbol: Krtap16-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183296.1 | NP_899119.1 | 1 | 507 | ||
GenBank mRNA | AF345293.1 | - | 1 | 447 | ||
AK017265.1 | - | 1 | 507 | |||
BC107303.2 | - | 1 | 443 | |||
BC107304.2 | - | 1 | 443 | |||
BC119503.1 | - | 1 | 290 | |||
BC119504.2 | - | 1 | 443 | |||
BC127231.1 | - | 1 | 443 | |||
BC127232.1 | - | 1 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
