siRNA Details
Gene Symbol: Cyp4f13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130882.1 | NP_570952.1 | 7 | 898 | ||
GenBank mRNA | AF233643.1 | - | 7 | 898 | ||
BC003954.1 | - | 7 | 907 | 3487830 | ||
BC058330.1 | - | 4 | 590 |
