siRNA Details
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130163.1 | NP_001123635.1 | 11 | 2034 | ||
NM_001130164.1 | NP_001123636.1 | 3 | 387 | |||
NM_001130165.1 | NP_001123637.1 | 11 | 2129 | |||
NM_001130166.1 | NP_001123638.1 | 14 | 2474 | |||
NM_130885.2 | NP_570955.1 | 12 | 2115 | |||
GenBank mRNA | AF324899.1 | - | 12 | 2114 | ||
AF333985.1 | - | 11 | 2034 | |||
AK158703.1 | - | 14 | 2456 | |||
AW106090.1 | - | 3 | 400 | |||
BC003927.1 | - | 5 | 624 | |||
BC089183.1 | - | 11 | 2124 | 5321255 | ||
BC098491.1 | - | 13 | 2376 | 30086857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
