siRNA Details
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130163.1 | NP_001123635.1 | 8 | 1595 | ||
NM_001130165.1 | NP_001123637.1 | 8 | 1690 | |||
NM_001130166.1 | NP_001123638.1 | 10 | 1954 | |||
NM_130885.2 | NP_570955.1 | 8 | 1595 | |||
GenBank mRNA | AF324899.1 | - | 8 | 1594 | ||
AF333985.1 | - | 8 | 1595 | |||
AK040881.1 | - | 8 | 1618 | |||
AK158703.1 | - | 10 | 1936 | |||
BC003927.1 | - | 2 | 185 | |||
BC089183.1 | - | 8 | 1685 | 5321255 | ||
BC098491.1 | - | 10 | 1937 | 30086857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
