siRNA Details
Gene Symbol: Scrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130893.3 | NP_570963.1 | 2 | 1367 | ||
GenBank mRNA | BC057562.1 | - | 2 | 1140 | 6401572 | |
BC089316.1 | - | 2 | 1149 |

More Information
Other Species Targeted

Gene Ontology Categories:
