siRNA Details
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285920.1 | NP_001272849.1 | 14 | 1820 | ||
NM_001285921.1 | NP_001272850.1 | 14 | 1803 | |||
NM_001285922.1 | NP_001272851.1 | 13 | 1734 | |||
NM_001285923.1 | NP_001272852.1 | 13 | 1734 | |||
NM_133201.3 | NP_573464.2 | 13 | 1737 | |||
GenBank mRNA | AB048831.1 | - | 11 | 1338 | ||
AF384100.1 | - | 14 | 1617 | |||
AK042080.1 | - | 13 | 1518 | |||
AK049583.1 | - | 13 | 1527 | |||
AK052689.1 | - | 13 | 1518 | |||
AK122220.1 | - | 12 | 2712 | |||
AK146801.1 | - | 14 | 1580 | |||
AK147863.1 | - | 13 | 1533 | |||
AK169373.1 | - | 13 | 1522 | |||
AY028170.1 | - | 1498 | ||||
AY123975.1 | - | 13 | 1684 | |||
BC046503.1 | - | 13 | 1515 | 4191179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
