siRNA Details
Gene Symbol: Pilrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133209.2 | NP_573472.2 | 2 | 561 | ||
GenBank mRNA | AB122024.2 | - | 2 | 456 | ||
AJ400847.1 | - | 2 | 510 | |||
BC145880.1 | - | 2 | 499 | 40131183 | ||
BC145906.1 | - | 2 | 499 | 40131209 | ||
BC172053.1 | - | 2 | 499 | 9054339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
