siRNA Details
Gene Symbol: Tlr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290755.1 | NP_001277684.1 | 5 | 1075 | ||
NM_001290756.1 | NP_001277685.1 | 4 | 994 | |||
NM_001290757.1 | NP_001277686.1 | 3 | 962 | |||
NM_001290758.1 | NP_001277687.1 | 4 | 1007 | |||
NM_133211.4 | NP_573474.1 | 3 | 926 | |||
GenBank mRNA | AF334942.1 | - | 3 | 869 | ||
AF334943.1 | - | 3 | 883 | |||
AK036549.1 | - | 3 | 884 | |||
AK154906.1 | - | 5 | 1033 | |||
AK156908.1 | - | 3 | 930 | |||
AK172680.1 | - | 1 | 4886 | |||
AY035889.1 | - | 2 | 743 | |||
BC120596.1 | - | 2 | 788 | 40129519 | ||
BC132385.1 | - | 3 | 856 | 40130662 |
