siRNA Details
Gene Symbol: Acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133225.3 | NP_573488.2 | 6 | 1050 | ||
GenBank mRNA | AF022770.3 | - | 6 | 1051 | ||
AK030371.1 | - | 6 | 1051 | |||
AK167272.1 | - | 6 | 1032 | |||
AK168799.1 | - | 6 | 1023 | |||
BC060602.1 | - | 6 | 1005 | 5706182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
