siRNA Details
Gene Symbol: Pdzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133226.2 | NP_573489.2 | 4 | 287 | ||
GenBank mRNA | AF334612.1 | - | 4 | 283 | ||
AK052748.1 | - | 4 | 288 |

More Information
Other Species Targeted

Gene Ontology Categories:
