siRNA Details
Gene Symbol: Bbc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133234.2 | NP_573497.1 | 4 | 776 | ||
GenBank mRNA | AF332560.1 | - | 4 | 721 | ||
BC044782.2 | - | 4 | 743 | |||
BC060370.1 | - | 4 | 706 | |||
BC147261.1 | - | 4 | 604 | 9056294 | ||
BC147262.1 | - | 4 | 604 | 9056313 |
