siRNA Details
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133241.2 | NP_573504.1 | 5 | 450 | ||
GenBank mRNA | AF449425.1 | - | 5 | 904 | ||
AK030396.1 | - | 5 | 449 | |||
AK043757.1 | - | 5 | 450 | |||
AK122190.1 | - | 5 | 892 | |||
BC024719.1 | - | 5 | 450 | 5363238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
