siRNA Details
Gene Symbol: Tram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177409.3 | NP_803128.1 | 4 | 456 | ||
GenBank mRNA | AK082751.1 | - | 4 | 457 | ||
AY029530.1 | - | 4 | 393 | |||
BC018212.1 | - | 4 | 441 | 3980004 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002730009.
Gene Ontology Categories:
