siRNA Details
Gene Symbol: Tram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177409.3 | NP_803128.1 | 4 | 510 | ||
GenBank mRNA | AK082751.1 | - | 4 | 511 | ||
AY029530.1 | - | 4 | 447 | |||
BC018212.1 | - | 4 | 495 | 3980004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
