siRNA Details
Gene Symbol: Zfp369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178364.5 | NP_848141.3 | 8 | 1386 | ||
GenBank mRNA | AJ319726.1 | - | 8 | 1403 | ||
AK084309.1 | - | 8 | 1389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
