siRNA Details
Gene Symbol: Zfp617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133358.3 | NP_579936.2 | 3, 4 | 344 | ||
GenBank mRNA | AB020542.1 | - | 305 | |||
AK143880.1 | - | 3, 4 | 318 | |||
BC014712.1 | - | 3, 4 | 250 | 4507398 |
