siRNA Details
Gene Symbol: Il17rc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134159.4 | NP_598920.3 | 7 | 842 | ||
NM_178942.1 | NP_849273.1 | 8 | 914 | |||
GenBank mRNA | AF458066.1 | - | 7 | 723 | ||
AK075634.1 | - | 4 | 332 | |||
BC004759.1 | - | 8 | 862 | 3154616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
