siRNA Details
Gene Symbol: Vmn1r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134174.2 | NP_598935.2 | 1 | 517 | ||
GenBank mRNA | BC138068.1 | - | 1 | 564 | 8861087 | |
BC138069.1 | - | 1 | 564 | 8861088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
