siRNA Details
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133643.4 | NP_598398.3 | 6 | 880 | ||
GenBank mRNA | AF358671.1 | - | 6 | 874 | ||
AK041545.1 | - | 6 | 814 | |||
AK042128.1 | - | 6 | 538 | |||
AK042414.1 | - | 6 | 843 | |||
AK075786.1 | - | 6 | 516 | |||
AK148351.1 | - | 6 | 702 | |||
AY071863.1 | - | 6 | 378 | |||
BC030683.1 | - | 6 | 456 | 1344937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
