siRNA Details
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133643.4 | NP_598398.3 | 6 | 860 | ||
GenBank mRNA | AF358671.1 | - | 6 | 854 | ||
AK041545.1 | - | 6 | 794 | |||
AK042128.1 | - | 6 | 518 | |||
AK042414.1 | - | 6 | 823 | |||
AK075786.1 | - | 6 | 496 | |||
AK148351.1 | - | 6 | 682 | |||
AY071863.1 | - | 6 | 358 | |||
BC030683.1 | - | 6 | 436 | 1344937 |
