siRNA Details
Gene Symbol: Galnt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134189.2 | NP_598950.2 | 3 | 459 | ||
GenBank mRNA | AK033515.1 | - | 2 | 117 | ||
AK131155.1 | - | 3 | 437 | |||
AK143714.1 | - | 3 | 457 | |||
AK159362.1 | - | 3 | 457 | |||
AK162424.1 | - | 3 | 459 | |||
AK172609.1 | - | 3 | 439 | |||
BB626609.1 | - | 3 | 459 | |||
BC060617.1 | - | 3 | 442 | 6836791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
