siRNA Details
Gene Symbol: Vmn1r226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134191.1 | NP_598952.1 | 1 | 166 | ||
GenBank mRNA | BC119429.1 | - | 1 | 164 | ||
BC119430.1 | - | 1 | 166 | 40040474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
