siRNA Details
Gene Symbol: Vmn1r232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134193.2 | NP_598954.2 | 1 | 328 | ||
GenBank mRNA | BC132631.1 | - | 1 | 328 | 40130908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
