siRNA Details
Gene Symbol: PCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000282.3 | NP_000273.2 | 2 | 245 | ||
NM_001178004.1 | NP_001171475.1 | 2 | 245 | |||
XM_005254059.2 | XP_005254116.1 | 2 | 240 | |||
XM_011521093.1 | XP_011519395.1 | 2 | 240 | |||
XM_017020605.1 | XP_016876094.1 | 2 | 240 | |||
XM_017020607.1 | XP_016876096.1 | 2 | 56 | |||
XM_017020608.1 | XP_016876097.1 | 2 | 240 | |||
XM_017020609.1 | XP_016876098.1 | 2 | 153 | |||
XM_017020611.1 | XP_016876100.1 | 2 | 240 | |||
XM_017020612.1 | XP_016876101.1 | 2 | 240 | |||
XM_017020613.1 | XP_016876102.1 | 2 | 240 | |||
XM_017020615.1 | XP_016876104.1 | 2 | 240 | |||
XM_017020616.1 | XP_016876105.1 | 2 | 240 | |||
GenBank mRNA | AF385926.1 | - | 2 | 177 | ||
AK225288.1 | - | 2 | 145 | |||
AK225299.1 | - | 2 | 143 | |||
AK298318.1 | - | 2 | 167 | |||
AK315503.1 | - | 2 | 143 | |||
BC000140.2 | - | 2 | 117 | 3048998 | ||
X14608.1 | - | 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
