siRNA Details
Gene Symbol: PCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000282.3 | NP_000273.2 | 10 | 874 | ||
NM_001127692.2 | NP_001121164.1 | 9 | 796 | |||
NM_001178004.1 | NP_001171475.1 | 10 | 874 | |||
XM_005254059.2 | XP_005254116.1 | 10 | 869 | |||
XM_011521093.1 | XP_011519395.1 | 10 | 869 | |||
XM_017020605.1 | XP_016876094.1 | 10 | 869 | |||
XM_017020606.1 | XP_016876095.1 | 9 | 720 | |||
XM_017020607.1 | XP_016876096.1 | 10 | 685 | |||
XM_017020608.1 | XP_016876097.1 | 10 | 869 | |||
XM_017020609.1 | XP_016876098.1 | 10 | 782 | |||
XM_017020610.1 | XP_016876099.1 | 8 | 1136 | |||
XM_017020611.1 | XP_016876100.1 | 10 | 869 | |||
XM_017020612.1 | XP_016876101.1 | 10 | 869 | |||
XM_017020613.1 | XP_016876102.1 | 10 | 869 | |||
XM_017020614.1 | XP_016876103.1 | 9 | 720 | |||
XM_017020615.1 | XP_016876104.1 | 10 | 869 | |||
XM_017020616.1 | XP_016876105.1 | 10 | 869 | |||
XM_017020617.1 | XP_016876106.1 | 6 | 453 | |||
XM_017020618.1 | XP_016876107.1 | 6 | 453 | |||
GenBank mRNA | AF385926.1 | - | 10 | 806 | ||
AK225288.1 | - | 10 | 774 | |||
AK225299.1 | - | 10 | 772 | |||
AK296771.1 | - | 9 | 694 | |||
AK298318.1 | - | 10 | 796 | |||
AK315503.1 | - | 10 | 772 | |||
BC000140.2 | - | 10 | 746 | 3048998 | ||
X14608.1 | - | 10 | 741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
