siRNA Details
Gene Symbol: Vmn1r67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134229.2 | NP_598990.2 | 1 | 875 | ||
GenBank mRNA | BC138174.1 | - | 1 | 875 | 8861193 | |
BC145742.1 | - | 1 | 875 | 40131045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
