siRNA Details
Gene Symbol: Vmn1r220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134237.1 | NP_598998.1 | 1 | 793 |
Gene Symbol: Vmn1r217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134239.1 | NP_599000.1 | 1 | 793 |
