siRNA Details
Gene Symbol: Vmn1r211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134243.1 | NP_599004.1 | 1 | 746 | ||
GenBank mRNA | BC138182.1 | - | 1 | 794 | 8861201 | |
BC138183.1 | - | 1 | 794 | 8861202 |
Gene Symbol: Vmn1r197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134244.1 | NP_599005.1 | 1 | 746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
